Learn More
Medchemexpress LLC Benzarone | 1477-19-6 | 99.7% | 266.29 | 500 MG
SDP
Catalog No. 5000453784
Encompass_Preferred
Benzarone is an oral inhibitor of human urate transporter 1 (hURAT1) with an IC50 value of 2.8 μM, and it also functions as an uncoupler of oxidative phosphorylation. This compound has the potential to cause liver damage and can promote cell apoptosis and necrosis. It is utilized to reduce serum uric acid levels and for research related to vascular diseases.
  • Oral inhibitor of human urate transporter 1 (hURAT1).
  • Functions as an uncoupler of oxidative phosphorylation.
  • Used to lower serum uric acid levels.
  • Useful for research in vascular diseases.
  • May cause liver damage and promote cell apoptosis and necrosis.
Catalog No. 50-004-53784 Supplier Medchemexpress LLC Supplier No. HYW011711500MG
Benzarone is an oral inhibitor of human urate transporter 1 (hURAT1) with an IC50 value of 2.8 μM, and it also functions as an uncoupler of oxidative phosphorylation. This compound has the potential to cause liver damage and can promote cell apoptosis and necrosis. It is utilized to reduce serum uric acid levels and for research related to vascular diseases.
  • Oral inhibitor of human urate transporter 1 (hURAT1).
  • Functions as an uncoupler of oxidative phosphorylation.
  • Used to lower serum uric acid levels.
  • Useful for research in vascular diseases.
  • May cause liver damage and promote cell apoptosis and necrosis.