Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzathine benzylpenicillin (tetrahydrate) | 41372-02-5 | 1 ML
SDP

Catalog No. 5000418074
Encompass_Preferred

Penicillin G benzathine tetrahydrate, also known as Benzathine benzylpenicillin tetrahydrate, is an antibiotic effective against a wide range of bacterial infections. It has a molecular weight of 981.18 and a chemical formula of C₄₈H₆₄N₆O₁₂S₂. This product is typically an off-white to light yellow solid, intended for research use only.

  • Effective against many bacterial infections
  • Molecular weight of 981.18
  • Chemical formula: C₄₈H₆₄N₆O₁₂S₂
  • Appearance: solid, off-white to light yellow
  • Purity: 99.02%
  • Soluble in DMSO for in vitro applications
  • Recommended storage as powder at -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-18074 Supplier Medchemexpress LLC Supplier No. HYN7139B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Penicillin G benzathine tetrahydrate, also known as Benzathine benzylpenicillin tetrahydrate, is an antibiotic effective against a wide range of bacterial infections. It has a molecular weight of 981.18 and a chemical formula of C₄₈H₆₄N₆O₁₂S₂. This product is typically an off-white to light yellow solid, intended for research use only.

  • Effective against many bacterial infections
  • Molecular weight of 981.18
  • Chemical formula: C₄₈H₆₄N₆O₁₂S₂
  • Appearance: solid, off-white to light yellow
  • Purity: 99.02%
  • Soluble in DMSO for in vitro applications
  • Recommended storage as powder at -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.