Learn More
Apexbio Technology LLC Benzbromarone 3562-84-3 50mg
SDP
Catalog No. 502240798
Encompass_Preferred
Benzbromarone (CAS 3562-84-3) is a small molecule inhibitor that selectively targets cytochrome P450 2C9 (CYP2C9) a key enzyme involved in the hepatic metabolism of various xenobiotics and endogenous compounds It interacts with CYP2C9 with a reported inhibition constant (K sub i /sub ) of 19 3 nM indicating high binding affinity Benzbromarone is utilized in biomedical research to study CYP2C9-mediated metabolic pathways and to evaluate drug-drug interactions that may arise from modulation of this enzyme s activity Its use facilitates investigations into hepatic metabolism pharmacokinetics and the enzymatic basis of adverse drug reactions
This item is not returnable. View return policy
Catalog No. 50-224-0798 Supplier Apexbio Technology LLC Supplier No. B167350
Benzbromarone (CAS 3562-84-3) is a small molecule inhibitor that selectively targets cytochrome P450 2C9 (CYP2C9) a key enzyme involved in the hepatic metabolism of various xenobiotics and endogenous compounds It interacts with CYP2C9 with a reported inhibition constant (K sub i /sub ) of 19 3 nM indicating high binding affinity Benzbromarone is utilized in biomedical research to study CYP2C9-mediated metabolic pathways and to evaluate drug-drug interactions that may arise from modulation of this enzyme s activity Its use facilitates investigations into hepatic metabolism pharmacokinetics and the enzymatic basis of adverse drug reactions