Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzbromarone, 95%, MilliporeSigma™ Supelco™

Catalog No. 111019819 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9819 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9819 Supplier MilliporeSigma Supelco Supplier No. B57741G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard for use in forensics, toxicology, and pharmaceutical applications.

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzbromarone
CAS 3562-84-3
Physical Form Neat
Linear Formula C17H12 Br2O3
Technique Liquid Chromatography
MDL Number MFCD00078962
Synonym 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethylbenzofuran
Type Analytical Standard
Molecular Weight (g/mol) 424.08
Quantity 1 g
Formula Weight 424.08
Percent Purity 95%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.