Learn More
Medchemexpress LLC Benzbromarone-d5 | 3562-84-3 | 99.4% | 5 MG
SDP
Catalog No. 5000412594
Encompass_Preferred
Benzbromarone-d5 is deuterium labeled Benzbromarone. It is for research use only. We do not sell to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development. Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12594 Supplier Medchemexpress LLC Supplier No. HYB1135S5MG
Benzbromarone-d5 is deuterium labeled Benzbromarone. It is for research use only. We do not sell to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development. Deuteration may affect pharmacokinetic and metabolic profiles of drugs.