Learn More
Medchemexpress LLC Benzbromarone-d5 | 3562-84-3 | 99.42% | 1 MG
SDP
Catalog No. 5000412565
Encompass_Preferred
Benzbromarone-d5 is a deuterium labeled Benzbromarone, for research use only. It serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements, like those in Benzbromarone-d5, have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-004-12565 Supplier Medchemexpress LLC Supplier No. HYB1135S1MG
Benzbromarone-d5 is a deuterium labeled Benzbromarone, for research use only. It serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements, like those in Benzbromarone-d5, have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications