Learn More
Medchemexpress LLC Benzenamine-13C6, 4-iodo | 233600-80-1 | 99.2% | 224.98 | 13C6H6IN | 10 MG
SDP
Catalog No. 5000297421
Encompass_Preferred
4-Iodoaniline-13C6 is the 13C6-labeled analogue of 4-iodoaniline provided as a stable isotope reference and research reagent. It serves as an internal standard and synthetic intermediate for analytical and preparative applications, particularly in studies related to methemoglobinemia, glycogen phosphorylase inhibitor synthesis, and liver or blood disease research. The labeled compound has formula 13C6H6IN, CAS 233600-80-1, and molecular weight 224.98 g/mol.
  • Stable 13C6 isotopic labeling for accurate mass spectrometry quantification.
  • High chemical purity suitable for analytical and synthetic use.
  • Supplied in milligram quantities appropriate for tracer and reference applications.
  • Compatible with LC-MS and GC-MS workflows as an internal standard.
  • Useful as a building block for organic synthesis and method development.
Catalog No. 50-002-97421 Supplier Medchemexpress LLC Supplier No. HYI0210S10MG
4-Iodoaniline-13C6 is the 13C6-labeled analogue of 4-iodoaniline provided as a stable isotope reference and research reagent. It serves as an internal standard and synthetic intermediate for analytical and preparative applications, particularly in studies related to methemoglobinemia, glycogen phosphorylase inhibitor synthesis, and liver or blood disease research. The labeled compound has formula 13C6H6IN, CAS 233600-80-1, and molecular weight 224.98 g/mol.
  • Stable 13C6 isotopic labeling for accurate mass spectrometry quantification.
  • High chemical purity suitable for analytical and synthetic use.
  • Supplied in milligram quantities appropriate for tracer and reference applications.
  • Compatible with LC-MS and GC-MS workflows as an internal standard.
  • Useful as a building block for organic synthesis and method development.