Learn More
Medchemexpress LLC Benzenamine, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- | 214360-73-3 | 98.0% | C12H18BNO2 | 10 G
SDP
Catalog No. 5000433609
Encompass_Preferred
4-Aminophenylboronic acid pinacol ester is a biochemical reagent for life science research. Also known as 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaboran-2yl)aniline, it serves as a heterocyclic building block in various synthetic processes. This compound is notably used in the synthesis of 3-aminoindazole-based multi-targeted receptor tyrosine kinase (RTK) inhibitors with anticancer activity and in creating roscovitine derivatives that act as dual inhibitors of cyclin-dependent kinases (CDKs) and casein kinase 1 (CK1).
  • Biochemical reagent for life science research.
  • Suitable as a biological material or organic compound.
  • Used in synthesis of 3-aminoindazole-based multi-targeted RTK inhibitors.
  • Demonstrates anticancer activity when used in RTK inhibitors.
  • Utilized in creation of roscovitine derivatives.
  • Functions as a heterocyclic building block for in vitro use.
Catalog No. 50-004-33609 Supplier Medchemexpress LLC Supplier No. HY7559710G
4-Aminophenylboronic acid pinacol ester is a biochemical reagent for life science research. Also known as 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaboran-2yl)aniline, it serves as a heterocyclic building block in various synthetic processes. This compound is notably used in the synthesis of 3-aminoindazole-based multi-targeted receptor tyrosine kinase (RTK) inhibitors with anticancer activity and in creating roscovitine derivatives that act as dual inhibitors of cyclin-dependent kinases (CDKs) and casein kinase 1 (CK1).
  • Biochemical reagent for life science research.
  • Suitable as a biological material or organic compound.
  • Used in synthesis of 3-aminoindazole-based multi-targeted RTK inhibitors.
  • Demonstrates anticancer activity when used in RTK inhibitors.
  • Utilized in creation of roscovitine derivatives.
  • Functions as a heterocyclic building block for in vitro use.