Learn More
Medchemexpress LLC Benzenamine, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- | 214360-73-3 | ≥98.0% | C12H18BNO2 | 25 G
SDP
Catalog No. 5000358578
Encompass_Preferred
4-Aminophenylboronic acid pinacol ester is a biochemical reagent that can be used as a biological material or organic compound for life science related research. As a heterocyclic building block, it has been used in the synthesis of 3-aminoindazole-based multi-targeted receptor tyrosine kinase (RTK) inhibitors with anticancer activity and roscovitine derivatives that are dual inhibitors of cyclin-dependent kinases (CDKs) and casein kinase 1 (CK1).
  • Biochemical reagent
  • Biological material or organic compound for life science related research
  • Heterocyclic building block
  • Used in the synthesis of 3-aminoindazole-based multi-targeted receptor tyrosine kinase (RTK) inhibitors with anticancer activity
  • Used in the synthesis of roscovitine derivatives that are dual inhibitors of cyclin-dependent kinases (CDKs) and casein kinase 1 (CK1)
Catalog No. 50-003-58578 Supplier Medchemexpress LLC Supplier No. 25GHY75597
4-Aminophenylboronic acid pinacol ester is a biochemical reagent that can be used as a biological material or organic compound for life science related research. As a heterocyclic building block, it has been used in the synthesis of 3-aminoindazole-based multi-targeted receptor tyrosine kinase (RTK) inhibitors with anticancer activity and roscovitine derivatives that are dual inhibitors of cyclin-dependent kinases (CDKs) and casein kinase 1 (CK1).
  • Biochemical reagent
  • Biological material or organic compound for life science related research
  • Heterocyclic building block
  • Used in the synthesis of 3-aminoindazole-based multi-targeted receptor tyrosine kinase (RTK) inhibitors with anticancer activity
  • Used in the synthesis of roscovitine derivatives that are dual inhibitors of cyclin-dependent kinases (CDKs) and casein kinase 1 (CK1)