Learn More
Medchemexpress LLC Benzenamine, 4,4',4'-methylidynetris- | 548-61-8 | ≥98.0% | 1 ML
SDP
Catalog No. 5000409814
Encompass_Preferred
Tris(4-aminophenyl)methane is a triphenylmethane dye and a weak HCV helicase inhibitor. It shows 30% inhibition at 100 μM against HCV helicase and can be used as an RNA preservative in biological samples.
  • Triphenylmethane dye
  • Weak HCV helicase inhibitor
  • Shows 30% inhibition at 100 μM against HCV helicase
  • Can be used as an RNA preservative in biological samples
  • Precipitates RNA in an aqueous solution
  • Can be applied in immunostaining assays and other histochemical methods
Catalog No. 50-004-09814 Supplier Medchemexpress LLC Supplier No. HYD030610MM/1ML
Tris(4-aminophenyl)methane is a triphenylmethane dye and a weak HCV helicase inhibitor. It shows 30% inhibition at 100 μM against HCV helicase and can be used as an RNA preservative in biological samples.
  • Triphenylmethane dye
  • Weak HCV helicase inhibitor
  • Shows 30% inhibition at 100 μM against HCV helicase
  • Can be used as an RNA preservative in biological samples
  • Precipitates RNA in an aqueous solution
  • Can be applied in immunostaining assays and other histochemical methods