Learn More
Medchemexpress LLC Benzenamine, 4,4'-carbonimidoylbis[N,N-dimethyl-, hydrochloride (1:1) | 2465-27-2 | ≥95.0% | 50 G
SDP
Catalog No. 5000410083
Encompass_Preferred
Auramine O is a yellow fluorescent dye used to detect amyloid fibrils, promotes lung malignancy, and is also used to determine algal cell viability and stain acid-fast bacteria.
  • Fluorescent dye
  • Detects amyloid fibrils
  • Promotes lung malignancy
  • Used to determine algal cell viability
  • Stains acid-fast bacteria
Catalog No. 50-004-10083 Supplier Medchemexpress LLC Supplier No. HYD093350G
Auramine O is a yellow fluorescent dye used to detect amyloid fibrils, promotes lung malignancy, and is also used to determine algal cell viability and stain acid-fast bacteria.
  • Fluorescent dye
  • Detects amyloid fibrils
  • Promotes lung malignancy
  • Used to determine algal cell viability
  • Stains acid-fast bacteria