Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenamine, 4-(8-methyl-9H-1,3-dioxolo[4,5-h]benzodiazepin-5-yl)-, hydrochloride (1:2) | 2319722-40-0 | 99.8% | 366.24 | C17H17Cl2N3O2 | 1 ML
SDP

Catalog No. 5000245955
Encompass_Preferred

GYKI 52466 dihydrochloride is a selective, noncompetitive AMPA/kainate receptor antagonist used in neuroscience research. It is orally active, shows good blood-brain barrier permeability, and has demonstrated anticonvulsant effects. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid quantities for experimental needs.

  • High purity, suitable for research applications (≈99.8%).
  • Available as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • Molecular formula C17H17Cl2N3O2, molecular weight 366.24.
  • Useful for studies of excitatory neurotransmission and neuroprotection.
  • Demonstrated blood-brain barrier permeability and anticonvulsant activity.
  • CAS number 2319722-40-0 for the dihydrochloride salt.

Catalog No. 50-002-45955 Supplier Medchemexpress LLC Supplier No. HY103234A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GYKI 52466 dihydrochloride is a selective, noncompetitive AMPA/kainate receptor antagonist used in neuroscience research. It is orally active, shows good blood-brain barrier permeability, and has demonstrated anticonvulsant effects. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid quantities for experimental needs.

  • High purity, suitable for research applications (≈99.8%).
  • Available as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • Molecular formula C17H17Cl2N3O2, molecular weight 366.24.
  • Useful for studies of excitatory neurotransmission and neuroprotection.
  • Demonstrated blood-brain barrier permeability and anticonvulsant activity.
  • CAS number 2319722-40-0 for the dihydrochloride salt.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.