Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenamine, 5-bromo-2-(diethylphosphinyl)-N-methyl- | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 g·mol⁻¹ | C11H17BrNOP | 10 G
SDP

Catalog No. 5000281753
Encompass_Preferred

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a drug intermediate used in medicinal chemistry and synthetic organic chemistry for the preparation of bioactive molecules. It is supplied as a high-purity solid for research and development applications; consult the certificate of analysis and safety data sheet before use.

  • High reported purity: 99.8%.
  • Molecular formula: C11H17BrNOP.
  • Molecular weight: 290.14 g·mol⁻¹.
  • Available in multiple pack sizes, including 10 G.
  • Supplied with COA and SDS for quality and safety information.
  • Suitable as a synthetic intermediate in medicinal chemistry research.

Catalog No. 50-002-81753 Supplier Medchemexpress LLC Supplier No. HYW44159210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a drug intermediate used in medicinal chemistry and synthetic organic chemistry for the preparation of bioactive molecules. It is supplied as a high-purity solid for research and development applications; consult the certificate of analysis and safety data sheet before use.

  • High reported purity: 99.8%.
  • Molecular formula: C11H17BrNOP.
  • Molecular weight: 290.14 g·mol⁻¹.
  • Available in multiple pack sizes, including 10 G.
  • Supplied with COA and SDS for quality and safety information.
  • Suitable as a synthetic intermediate in medicinal chemistry research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.