Learn More
Medchemexpress LLC Benzenamine, N,N-bis(2-chloroethyl)- | 553-27-5 | 99.4% | 5 G
SDP
Catalog No. 5000231517
Encompass_Preferred
Aniline mustard (N,N-bis(2-chloroethyl)aniline) is an alkylating anticancer research reagent used as a cytotoxic agent in pharmacology and in vivo studies. It is supplied as a solid and as a DMSO solution, with documented solubility, storage, and handling recommendations for research applications.
  • Alkylating agent used as an anticancer research reagent.
  • High purity for research use (99.36%).
  • Formula C10H13Cl2N; molecular weight 218.13 g/mol.
  • Supplied as solid and DMSO solution with multiple pack sizes.
  • Store solid at 4°C and protect from light; follow solvent storage guidelines.
Catalog No. 50-002-31517 Supplier Medchemexpress LLC Supplier No. HY1217135G
Aniline mustard (N,N-bis(2-chloroethyl)aniline) is an alkylating anticancer research reagent used as a cytotoxic agent in pharmacology and in vivo studies. It is supplied as a solid and as a DMSO solution, with documented solubility, storage, and handling recommendations for research applications.
  • Alkylating agent used as an anticancer research reagent.
  • High purity for research use (99.36%).
  • Formula C10H13Cl2N; molecular weight 218.13 g/mol.
  • Supplied as solid and DMSO solution with multiple pack sizes.
  • Store solid at 4°C and protect from light; follow solvent storage guidelines.