Learn More
Medchemexpress LLC Benzene, 1,1'-(2,2-dichloroethylidene)bis[4-chloro- | 72-54-8 | 99.4% | 1 ML
SDP
Catalog No. 5000411272
Encompass_Preferred
p,p'-DDD is an organochlorine insecticide and a major metabolite of p,p'-DDT. It acts as an agonist at estrogen receptor α (ERα) and ERβ, and increases DNA damage, apoptosis, and necrosis in peripheral blood. It stimulates cell proliferation in SKBR3 cells, activates the AP-1 transcription factor, and has been shown to decrease sleep times of barbiturates and steroids in rats.
  • Organochlorine insecticide
  • Metabolite of p,p'-DDT
  • Agonist at estrogen receptor α and ERβ
  • Increases DNA damage, apoptosis, and necrosis in peripheral blood
  • Stimulates cell proliferation in SKBR3 cells
  • Activates AP-1 transcription factor
  • Decreases sleep times of barbiturates and steroids in rats
Catalog No. 50-004-11272 Supplier Medchemexpress LLC Supplier No. HYB198410MM/1ML
p,p'-DDD is an organochlorine insecticide and a major metabolite of p,p'-DDT. It acts as an agonist at estrogen receptor α (ERα) and ERβ, and increases DNA damage, apoptosis, and necrosis in peripheral blood. It stimulates cell proliferation in SKBR3 cells, activates the AP-1 transcription factor, and has been shown to decrease sleep times of barbiturates and steroids in rats.
  • Organochlorine insecticide
  • Metabolite of p,p'-DDT
  • Agonist at estrogen receptor α and ERβ
  • Increases DNA damage, apoptosis, and necrosis in peripheral blood
  • Stimulates cell proliferation in SKBR3 cells
  • Activates AP-1 transcription factor
  • Decreases sleep times of barbiturates and steroids in rats