Learn More
Medchemexpress LLC Benzene-1,3,5-tricarboxylic acid | 554-95-0 | MFCD00002517 | 99.0% | 210.14 g/mol | C9H6O6 | 1 ML
SDP
Catalog No. 5000298387
Encompass_Preferred
Benzene-1,3,5-tricarboxylic acid (trimesic acid) supplied as a 10 mM solution in DMSO (1 mL). It is an aromatic tricarboxylic scaffold commonly used in biochemical and materials research for peptide/protein binding studies and hydrogel stabilization.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity (98.96%).
  • Molecular weight 210.14 g/mol; formula C9H6O6.
  • Useful as a scaffold for peptide and protein binding studies.
  • Improves mechanical stability of hydrogels.
Catalog No. 50-002-98387 Supplier Medchemexpress LLC Supplier No. HY78985S10MG
Benzene-1,3,5-tricarboxylic acid (trimesic acid) supplied as a 10 mM solution in DMSO (1 mL). It is an aromatic tricarboxylic scaffold commonly used in biochemical and materials research for peptide/protein binding studies and hydrogel stabilization.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity (98.96%).
  • Molecular weight 210.14 g/mol; formula C9H6O6.
  • Useful as a scaffold for peptide and protein binding studies.
  • Improves mechanical stability of hydrogels.