Learn More
Medchemexpress LLC Benzene-1,3,5-tricarboxylic acid-d3 (Trimesic acid-d3) | 62790-27-6 | 99.2% | C9H3D3O6 | 5 MG
SDP
Catalog No. 5000433005
Encompass_Preferred
Benzene-1,3,5-tricarboxylic acid-d3 is a deuterium labeled form of Benzene-1,3,5-tricarboxylic acid. This scaffold enhances the binding ability of bicyclic peptides to proteins and improves the mechanical stability of hydrogels. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-33005 Supplier Medchemexpress LLC Supplier No. HY78985S5MG
Benzene-1,3,5-tricarboxylic acid-d3 is a deuterium labeled form of Benzene-1,3,5-tricarboxylic acid. This scaffold enhances the binding ability of bicyclic peptides to proteins and improves the mechanical stability of hydrogels. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.