Learn More
Medchemexpress LLC Benzene-1,3,5-tricarboxylic acid (Trimesic acid) | 554-95-0 | 99.8% | C9H6O6 | 50 G
SDP
Catalog No. 5000434903
Encompass_Preferred
Benzene-1,3,5-tricarboxylic acid, also known as Trimesic acid, functions as a scaffold in various applications. It is for research use only.
  • Enhances the binding ability of bicyclic peptides to proteins.
  • Improves the mechanical stability of hydrogels.
Catalog No. 50-004-34903 Supplier Medchemexpress LLC Supplier No. HY7898550G
Benzene-1,3,5-tricarboxylic acid, also known as Trimesic acid, functions as a scaffold in various applications. It is for research use only.
  • Enhances the binding ability of bicyclic peptides to proteins.
  • Improves the mechanical stability of hydrogels.