Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzene, 1,3-di(methoxy-d3)- | 16469-85-5 | 976500 | 98.0% | 144.20 | 50 MG
SDP

Catalog No. 5000404251
Encompass_Preferred

MedChemExpress 1,3-Dimethoxybenzene-d6 (HY-34487S2) is a deuterated chemical compound with a molecular weight of 144.20. It appears as a light yellow to yellow liquid and has a purity of 98.0% by GC. This product is intended for research use only and is not fully validated for medical applications.

  • Appearance: Light yellow to yellow liquid
  • Purity (GC): 98.0%
  • Isotopic enrichment: 99.9%
  • Storage (pure form): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-04251 Supplier Medchemexpress LLC Supplier No. HY34487S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MedChemExpress 1,3-Dimethoxybenzene-d6 (HY-34487S2) is a deuterated chemical compound with a molecular weight of 144.20. It appears as a light yellow to yellow liquid and has a purity of 98.0% by GC. This product is intended for research use only and is not fully validated for medical applications.

  • Appearance: Light yellow to yellow liquid
  • Purity (GC): 98.0%
  • Isotopic enrichment: 99.9%
  • Storage (pure form): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.