Learn More
Medchemexpress LLC Benzene, 1,4-dimethoxy-d10 | 74079-00-8 | 99.6% | 148.23 g·mol⁻1 | C8D10O2 | 10 MG
SDP
Catalog No. 5000300299
Encompass_Preferred
1,4-Dimethoxybenzene-d10 is the deuterium-labeled isotopologue of 1,4-dimethoxybenzene supplied for analytical and research use as a tracer or internal standard. It is characterized by high isotopic incorporation and is intended for quantitative and qualitative analyses where a mass- or NMR-resolved labelled analog is required.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic enrichment (~99.6% atom D) for reliable tracing.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in multiple small-scale packaging sizes for analytical work.
  • Provided with batch-specific certificate of analysis for traceability.
Catalog No. 50-003-00299 Supplier Medchemexpress LLC Supplier No. HYW015780S10MG
1,4-Dimethoxybenzene-d10 is the deuterium-labeled isotopologue of 1,4-dimethoxybenzene supplied for analytical and research use as a tracer or internal standard. It is characterized by high isotopic incorporation and is intended for quantitative and qualitative analyses where a mass- or NMR-resolved labelled analog is required.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic enrichment (~99.6% atom D) for reliable tracing.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in multiple small-scale packaging sizes for analytical work.
  • Provided with batch-specific certificate of analysis for traceability.