Learn More
Medchemexpress LLC Benzene-2,3,4,5-d4-butanoic acid, α,6-diamino-γ-oxo-, (S)- (hydrochloride) | 00-00-0 | 99.6% | 248.70 | C10H9D4ClN2O3 | 1 MG
SDP
Catalog No. 5000241881
Encompass_Preferred
L-Kynurenine-d4-1 hydrochloride is a deuterium-labeled analog of L-kynurenine supplied as the hydrochloride salt for research use. It is intended as a tracer and a stable internal standard for quantitative method development and metabolite analysis in biological samples.
Catalog No. 50-002-41881 Supplier Medchemexpress LLC Supplier No. HY104026S21MG
L-Kynurenine-d4-1 hydrochloride is a deuterium-labeled analog of L-kynurenine supplied as the hydrochloride salt for research use. It is intended as a tracer and a stable internal standard for quantitative method development and metabolite analysis in biological samples.