Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzene-d5, 6,6′-thiobis- | 180802-01-1 | 99.5% | 196.33 g·mol⁻¹ | C12D10S | 25 MG
SDP

Catalog No. 5000304042
Encompass_Preferred

Diphenylsulfane-d1 is the deuterium-labeled analogue of diphenylsulfane supplied as a stable-isotope standard for analytical, tracer, and research applications. It is a colorless to light yellow liquid (C12D10S, 196.33 g·mol⁻¹) characterized by high chemical purity and isotopic enrichment; batch-specific COA and SDS are available.

  • High chemical purity: 99.5% (GC).
  • High isotopic enrichment: 99.7%.
  • Molecular weight 196.33 g·mol⁻¹.
  • Chemical formula C12D10S.
  • Colorless to light yellow liquid appearance.
  • Suitable as a stable-isotope analytical standard and tracer.
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months).

Catalog No. 50-003-04042 Supplier Medchemexpress LLC Supplier No. 25MGHYW016174S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenylsulfane-d1 is the deuterium-labeled analogue of diphenylsulfane supplied as a stable-isotope standard for analytical, tracer, and research applications. It is a colorless to light yellow liquid (C12D10S, 196.33 g·mol⁻¹) characterized by high chemical purity and isotopic enrichment; batch-specific COA and SDS are available.

  • High chemical purity: 99.5% (GC).
  • High isotopic enrichment: 99.7%.
  • Molecular weight 196.33 g·mol⁻¹.
  • Chemical formula C12D10S.
  • Colorless to light yellow liquid appearance.
  • Suitable as a stable-isotope analytical standard and tracer.
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.