Learn More
Medchemexpress LLC Benzene, (ethoxymethyl)- | 539-30-0 | MFCD00039903 | 98.3% | 136.19 g/mol | C9H12O | 1 G
SDP
Catalog No. 5000300624
Encompass_Preferred
(Ethoxymethyl)benzene (CAS 539-30-0) is an analytical reference standard and endogenous metabolite supplied as a liquid for research and analytical applications. It is characterized by molecular formula C9H12O, molecular weight 136.19 g/mol, and a typical HPLC purity of 98.27%.
  • High purity (98.3% by HPLC) suitable for analytical workflows.
  • Liquid, colorless to light yellow appearance.
  • Molecular formula C9H12O and molecular weight 136.19 g/mol.
  • Suitable as a reference standard for metabolite identification and method development.
  • Certificate of analysis provided with HPLC assay data.
Catalog No. 50-003-00624 Supplier Medchemexpress LLC Supplier No. HYW0176131G
(Ethoxymethyl)benzene (CAS 539-30-0) is an analytical reference standard and endogenous metabolite supplied as a liquid for research and analytical applications. It is characterized by molecular formula C9H12O, molecular weight 136.19 g/mol, and a typical HPLC purity of 98.27%.
  • High purity (98.3% by HPLC) suitable for analytical workflows.
  • Liquid, colorless to light yellow appearance.
  • Molecular formula C9H12O and molecular weight 136.19 g/mol.
  • Suitable as a reference standard for metabolite identification and method development.
  • Certificate of analysis provided with HPLC assay data.