Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzene, (ethoxymethyl)- | 539-30-0 | MFCD00039903 | 99.0% | 136.19 g/mol | C9H12O | 10 G
SDP

Catalog No. 5000300395
Encompass_Preferred

Benzene, (ethoxymethyl)- (CAS 539-30-0) is an endogenous metabolite supplied as a high-purity analytical standard for research and analytical workflows. It is commonly used as a reference standard in chromatographic and mass spectrometric analyses and as a reagent in chemical research.

  • High purity for analytical applications (~99%).
  • Liquid, colorless to light yellow, density ~0.949 g/cm3.
  • Molecular weight 136.19 g/mol; formula C9H12O.
  • Supplied with certificate of analysis and datasheet for quality assurance.
  • Recommended storage conditions provided to preserve stability.

Catalog No. 50-003-00395 Supplier Medchemexpress LLC Supplier No. HYW01761310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzene, (ethoxymethyl)- (CAS 539-30-0) is an endogenous metabolite supplied as a high-purity analytical standard for research and analytical workflows. It is commonly used as a reference standard in chromatographic and mass spectrometric analyses and as a reagent in chemical research.

  • High purity for analytical applications (~99%).
  • Liquid, colorless to light yellow, density ~0.949 g/cm3.
  • Molecular weight 136.19 g/mol; formula C9H12O.
  • Supplied with certificate of analysis and datasheet for quality assurance.
  • Recommended storage conditions provided to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.