Learn More
Medchemexpress LLC Benzene, (ethoxymethyl)- | 539-30-0 | MFCD00039903 | 99.0% | 136.19 g/mol | C9H12O | 5 G
SDP
Catalog No. 5000300427
Encompass_Preferred
(Ethoxymethyl)benzene is supplied as an analytical reference standard intended for research and analytical applications. It is an endogenous metabolite provided as a liquid with characterized physical properties and high purity for use in qualitative and quantitative assays.
  • Analytical reference standard for qualitative and quantitative analysis.
  • Suitable for HPLC, GC, and MS applications.
  • High purity for reliable assay and calibration results.
  • Provided as a liquid with known density for accurate dosing.
  • Available in multiple laboratory-friendly pack sizes.
Catalog No. 50-003-00427 Supplier Medchemexpress LLC Supplier No. HYW0176135G
(Ethoxymethyl)benzene is supplied as an analytical reference standard intended for research and analytical applications. It is an endogenous metabolite provided as a liquid with characterized physical properties and high purity for use in qualitative and quantitative assays.
  • Analytical reference standard for qualitative and quantitative analysis.
  • Suitable for HPLC, GC, and MS applications.
  • High purity for reliable assay and calibration results.
  • Provided as a liquid with known density for accurate dosing.
  • Available in multiple laboratory-friendly pack sizes.