Learn More
Apexbio Technology LLC 1,2,3,4,5,6-Hexabromocyclohexane(Synonyms: Hexabromocyclohexane, Benzene hexabromide, Hexabenzene bromide, HCH hexabromide), 10mM (in 1mL DMSO), CAS: 1837-91-8.
SDP
Catalog No. 5000060963
Encompass_Preferred
1 2 3 4 5 6-Hexabromocyclohexane (CAS 1837-91-8) is a small molecule inhibitor targeting Janus kinase 2 (JAK2) It directly suppresses JAK2 tyrosine kinase activity by blocking ligand-induced autophosphorylation Experimental data indicate that treatment with 1 M of the compound for 16 hours decreases JAK2 autophosphorylation by approximately 50% whereas exposure to 50 M essentially abolishes kinase activity Notably it does not exhibit cytotoxicity at concentrations up to 100 M This compound serves as a valuable research tool for studying JAK2 signaling pathways and related cellular processes
Catalog No. 50-000-60963 Supplier Apexbio Technology LLC Supplier No. A45105.1
1 2 3 4 5 6-Hexabromocyclohexane (CAS 1837-91-8) is a small molecule inhibitor targeting Janus kinase 2 (JAK2) It directly suppresses JAK2 tyrosine kinase activity by blocking ligand-induced autophosphorylation Experimental data indicate that treatment with 1 M of the compound for 16 hours decreases JAK2 autophosphorylation by approximately 50% whereas exposure to 50 M essentially abolishes kinase activity Notably it does not exhibit cytotoxicity at concentrations up to 100 M This compound serves as a valuable research tool for studying JAK2 signaling pathways and related cellular processes