Learn More
Medchemexpress LLC Benzeneacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.16 g/mol | C8H9NO | 5 G
SDP
Catalog No. 5000301651
Encompass_Preferred
2-Phenylacetamide (benzeneacetamide) is a small-molecule research chemical (CAS 103-81-1) used as a biochemical reagent and synthetic intermediate. It has been reported to modulate p38 MAPK signaling and has been studied for anti-inflammatory, antioxidant, and anti-fibrotic effects. The substance is supplied as a solid powder at high purity and should be stored according to supplier guidance to maintain stability.
  • High purity (≈99.9%).
  • Molecular formula C8H9NO; molar mass ~135.16 g/mol.
  • Solid powder form suitable for laboratory use.
  • Available in small laboratory pack sizes for research applications.
  • Reported biological activity: p38 MAPK pathway modulation and anti-inflammatory effects.
  • Stable under recommended storage conditions (refrigerated or frozen as advised).
Catalog No. 50-003-01651 Supplier Medchemexpress LLC Supplier No. HYW0181975G
Add to Cart
Add to Cart
2-Phenylacetamide (benzeneacetamide) is a small-molecule research chemical (CAS 103-81-1) used as a biochemical reagent and synthetic intermediate. It has been reported to modulate p38 MAPK signaling and has been studied for anti-inflammatory, antioxidant, and anti-fibrotic effects. The substance is supplied as a solid powder at high purity and should be stored according to supplier guidance to maintain stability.
  • High purity (≈99.9%).
  • Molecular formula C8H9NO; molar mass ~135.16 g/mol.
  • Solid powder form suitable for laboratory use.
  • Available in small laboratory pack sizes for research applications.
  • Reported biological activity: p38 MAPK pathway modulation and anti-inflammatory effects.
  • Stable under recommended storage conditions (refrigerated or frozen as advised).