Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 | C8H9NO | 1 G
SDP

Catalog No. 5000302044
Encompass_Preferred

2-Phenylacetamide (benzeneacetamide; CAS 103-81-1) is a small organic compound used as a biochemical reagent and synthetic intermediate. It is supplied as a white to off-white solid with high reported purity, suitable for research applications.

  • CAS number: 103-81-1.
  • Chemical formula: C8H9NO.
  • Molecular weight: 135.17.
  • Appearance: white to off-white solid; reported purity 99.9%.
  • Common applications: biochemical research, synthetic intermediate, pharmacology studies.
  • Available pack sizes: 10 mM solution in DMSO and solids from 500 mg to >100 g.

Catalog No. 50-003-02044 Supplier Medchemexpress LLC Supplier No. HYW0181971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylacetamide (benzeneacetamide; CAS 103-81-1) is a small organic compound used as a biochemical reagent and synthetic intermediate. It is supplied as a white to off-white solid with high reported purity, suitable for research applications.

  • CAS number: 103-81-1.
  • Chemical formula: C8H9NO.
  • Molecular weight: 135.17.
  • Appearance: white to off-white solid; reported purity 99.9%.
  • Common applications: biochemical research, synthetic intermediate, pharmacology studies.
  • Available pack sizes: 10 mM solution in DMSO and solids from 500 mg to >100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.