Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 | C8H9NO | 500 G
SDP

Catalog No. 5000302045
Encompass_Preferred

2-Phenylacetamide is a research-grade small molecule reported to inhibit the p38 MAPK signaling pathway and to exhibit estrogen-like, anti-inflammatory, antioxidant, anti-hypertensive, and anti-fibrotic activities. It is orally active and supplied as a powder for biochemical and pharmacological research.

  • Purity: 99.92%.
  • Molecular formula: C8H9NO.
  • Molecular weight: 135.17 g/mol.
  • Physical form: powder, suitable for dissolution in common laboratory solvents.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: biochemical and pharmacological research applications.

Catalog No. 50-003-02045 Supplier Medchemexpress LLC Supplier No. HYW018197500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylacetamide is a research-grade small molecule reported to inhibit the p38 MAPK signaling pathway and to exhibit estrogen-like, anti-inflammatory, antioxidant, anti-hypertensive, and anti-fibrotic activities. It is orally active and supplied as a powder for biochemical and pharmacological research.

  • Purity: 99.92%.
  • Molecular formula: C8H9NO.
  • Molecular weight: 135.17 g/mol.
  • Physical form: powder, suitable for dissolution in common laboratory solvents.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: biochemical and pharmacological research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.