Learn More
Medchemexpress LLC Benzeneacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 g/mol | C8H9NO | 100 G
SDP
Catalog No. 5000300805
Encompass_Preferred
2-Phenylacetamide (CAS 103-81-1) is a small-molecule research compound reported to inhibit p38 MAPK signaling and to have anti-inflammatory, antioxidant, anti-hypertensive, and anti-fibrosis activities. It is supplied as a solid and as a DMSO solution for laboratory use; high purity and batch-specific certificates of analysis support experimental reproducibility.
Catalog No. 50-003-00805 Supplier Medchemexpress LLC Supplier No. HYW018197100G
2-Phenylacetamide (CAS 103-81-1) is a small-molecule research compound reported to inhibit p38 MAPK signaling and to have anti-inflammatory, antioxidant, anti-hypertensive, and anti-fibrosis activities. It is supplied as a solid and as a DMSO solution for laboratory use; high purity and batch-specific certificates of analysis support experimental reproducibility.