Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 g/mol | C8H9NO | 50 G
SDP

Catalog No. 5000301433
Encompass_Preferred

2-Phenylacetamide (benzeneacetamide, CAS 103-81-1) is a small-molecule research chemical supplied as a white to off-white solid with reported HPLC purity of 99.92%. It is used in biological studies, including investigations of the p38 MAPK signaling pathway; for research use only.

  • White to off-white solid.
  • High purity (99.92% by HPLC).
  • Molecular formula C8H9NO; molecular weight 135.17 g/mol.
  • Reported activity against the p38 MAPK signaling pathway in research studies.
  • Storage: powder stable at -20°C for 3 years and at 4°C for 2 years.
  • Available in multiple laboratory pack sizes.

Catalog No. 50-003-01433 Supplier Medchemexpress LLC Supplier No. HYW01819750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylacetamide (benzeneacetamide, CAS 103-81-1) is a small-molecule research chemical supplied as a white to off-white solid with reported HPLC purity of 99.92%. It is used in biological studies, including investigations of the p38 MAPK signaling pathway; for research use only.

  • White to off-white solid.
  • High purity (99.92% by HPLC).
  • Molecular formula C8H9NO; molecular weight 135.17 g/mol.
  • Reported activity against the p38 MAPK signaling pathway in research studies.
  • Storage: powder stable at -20°C for 3 years and at 4°C for 2 years.
  • Available in multiple laboratory pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.