Learn More
Medchemexpress LLC Benzeneacetamide, 4-butoxy-N-hydroxy- | 2438-72-4 | 98.0% | 500 MG
SDP
Catalog No. 5000409283
Encompass_Preferred
Bufexamac is a selective IIb HDAC (HDAC6, HDAC10) and LTA4H dual inhibitor, demonstrating Kds of 0.53 μM for HDAC6 and 0.22 μM for HDAC10. It also functions as a nonsteroidal anti-inflammatory drug, useful in various research applications.
  • Selective dual inhibitor of HDAC6, HDAC10, and LTA4H.
  • Functions as a nonsteroidal anti-inflammatory drug.
  • Inhibits IFN-α secretion in peripheral blood mononuclear cells.
  • Inhibits HDAC10 to block autophagic flux, enhancing tumor-specific toxicity of DNA damage-inducing drugs.
  • Inhibits neutrophil chemotaxis via inhibition of LTA4H-mediated endogenous LTB4 biosynthesis.
  • Ameliorates LPS-induced acute lung injury in mice by targeting LTA4H.
Catalog No. 50-004-09283 Supplier Medchemexpress LLC Supplier No. HYB0494500MG
Bufexamac is a selective IIb HDAC (HDAC6, HDAC10) and LTA4H dual inhibitor, demonstrating Kds of 0.53 μM for HDAC6 and 0.22 μM for HDAC10. It also functions as a nonsteroidal anti-inflammatory drug, useful in various research applications.
  • Selective dual inhibitor of HDAC6, HDAC10, and LTA4H.
  • Functions as a nonsteroidal anti-inflammatory drug.
  • Inhibits IFN-α secretion in peripheral blood mononuclear cells.
  • Inhibits HDAC10 to block autophagic flux, enhancing tumor-specific toxicity of DNA damage-inducing drugs.
  • Inhibits neutrophil chemotaxis via inhibition of LTA4H-mediated endogenous LTB4 biosynthesis.
  • Ameliorates LPS-induced acute lung injury in mice by targeting LTA4H.