Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetamide, N-[2,3-dihydro-1-(phenylmethyl)-1H-pyrrolo[2,3-b]quinolin-4-yl] | 365565-02-2 | 99.3% | 10 MG
SDP

Catalog No. 5000231873
Encompass_Preferred

PGP-4008 is a small-molecule research compound that selectively inhibits P-glycoprotein (Pgp) and is used to study Pgp-mediated multidrug resistance in cellular and in vivo models. It is supplied as solid material and as a DMSO solution; reported molecular formula is C26H23N3O, and molecular weight is 393.48 g/mol.

  • Primary activity: P-glycoprotein inhibitor.
  • High purity: 99.3% (LCMS).
  • Molecular formula: C26H23N3O.
  • Molecular weight: 393.48 g/mol.
  • CAS number: 365565-02-2.
  • Available forms: solid (500 μg-100 mg) and 10 mM solution in DMSO.
  • Recommended storage: -20°C, protect from moisture.

Catalog No. 50-002-31873 Supplier Medchemexpress LLC Supplier No. HY11982310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGP-4008 is a small-molecule research compound that selectively inhibits P-glycoprotein (Pgp) and is used to study Pgp-mediated multidrug resistance in cellular and in vivo models. It is supplied as solid material and as a DMSO solution; reported molecular formula is C26H23N3O, and molecular weight is 393.48 g/mol.

  • Primary activity: P-glycoprotein inhibitor.
  • High purity: 99.3% (LCMS).
  • Molecular formula: C26H23N3O.
  • Molecular weight: 393.48 g/mol.
  • CAS number: 365565-02-2.
  • Available forms: solid (500 μg-100 mg) and 10 mM solution in DMSO.
  • Recommended storage: -20°C, protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.