Learn More
Medchemexpress LLC Benzeneacetamide, N-[3-[(3,4-dihydro-2,4-dioxo-3-propylthieno[3,2-d] | 2098776-03-3 | 98.42% | C24H23N3O3S | 50 MG
SDP
Catalog No. 5000355249
Encompass_Preferred
FEN1-IN-SC13 is a potent DNA fragmentation endonuclease 1 (FEN1) inhibitor with antitumor activity. It interferes with DNA replication and repair both in vitro and in cells.
  • Potent DNA fragmentation endonuclease 1 (FEN1) inhibitor.
  • Exhibits antitumor activity.
  • Interferes with DNA replication and repair in vitro and in cells.
Catalog No. 50-003-55249 Supplier Medchemexpress LLC Supplier No. HY14575850MG
FEN1-IN-SC13 is a potent DNA fragmentation endonuclease 1 (FEN1) inhibitor with antitumor activity. It interferes with DNA replication and repair both in vitro and in cells.
  • Potent DNA fragmentation endonuclease 1 (FEN1) inhibitor.
  • Exhibits antitumor activity.
  • Interferes with DNA replication and repair in vitro and in cells.