Learn More
Medchemexpress LLC Benzeneacetamide, N-(aminoiminomethyl)-2,6-dichloro-, hydrochloride (1:1) | 29110-48-3 | MFCD00798230 | 99.9% | 282.55 | 100MG
SDP
Catalog No. 5000383968
Encompass_Preferred
Guanfacine hydrochloride is an orally active and blood-brain barrier permeable noradrenergic α2A agonist that exhibits high selectivity for the α2A receptor subtype. It has demonstrated effects in producing hypotension and sedation. This compound can be utilized for research into various prefrontal cortex (PFC) cognitive disorders, such as Tourette's syndrome and attention deficit hyperactivity disorder (ADHD).
  • Orally active
  • Blood-brain barrier permeable
  • Noradrenergic α2A agonist
  • High selectivity for the α2A receptor subtype
  • Produces hypotension and sedation
  • Used for research into prefrontal cortex (PFC) cognitive disorders
  • Potentially useful for Tourette's syndrome
  • Potentially useful for attention deficit hyperactivity disorder (ADHD)
Catalog No. 50-003-83968 Supplier Medchemexpress LLC Supplier No. HY17416100MG
Guanfacine hydrochloride is an orally active and blood-brain barrier permeable noradrenergic α2A agonist that exhibits high selectivity for the α2A receptor subtype. It has demonstrated effects in producing hypotension and sedation. This compound can be utilized for research into various prefrontal cortex (PFC) cognitive disorders, such as Tourette's syndrome and attention deficit hyperactivity disorder (ADHD).
  • Orally active
  • Blood-brain barrier permeable
  • Noradrenergic α2A agonist
  • High selectivity for the α2A receptor subtype
  • Produces hypotension and sedation
  • Used for research into prefrontal cortex (PFC) cognitive disorders
  • Potentially useful for Tourette's syndrome
  • Potentially useful for attention deficit hyperactivity disorder (ADHD)