Learn More
Medchemexpress LLC Benzeneacetic acid, 3-methyl- | 621-36-3 | MFCD00004340 | 99.9% | 150.18 | C9H10O2 | 25 G
SDP
Catalog No. 5000209738
Encompass_Preferred
m-Tolylacetic acid (3-methylphenylacetic acid) is a substituted phenylacetic acid provided as a high-purity analytical standard and synthetic intermediate for organic synthesis and analytical chemistry. It is typically supplied as a white to off-white solid for research and analytical applications.
  • High purity standard (HPLC purity ~99.9%).
  • White to off-white solid physical form.
  • Molecular formula C9H10O2, molecular weight 150.18 g/mol.
  • Used as an analytical standard and synthetic intermediate in organic synthesis.
  • Supplied in 25 G packages (verify packaging with supplier).
Catalog No. 50-002-09738 Supplier Medchemexpress LLC Supplier No. 25GHYW053507
m-Tolylacetic acid (3-methylphenylacetic acid) is a substituted phenylacetic acid provided as a high-purity analytical standard and synthetic intermediate for organic synthesis and analytical chemistry. It is typically supplied as a white to off-white solid for research and analytical applications.
  • High purity standard (HPLC purity ~99.9%).
  • White to off-white solid physical form.
  • Molecular formula C9H10O2, molecular weight 150.18 g/mol.
  • Used as an analytical standard and synthetic intermediate in organic synthesis.
  • Supplied in 25 G packages (verify packaging with supplier).