Learn More
Medchemexpress LLC Benzeneacetic acid, alpha-hydroxy-, ethyl ester | 774-40-3 | MFCD00004494 | 99.7% | 180.20 g·mol-1 | C10H12O3 | 25 G
SDP
Catalog No. 5000303523
Encompass_Preferred
Ethyl mandelate is the ethyl ester of mandelic acid used as a pharmaceutical intermediate and building block in organic synthesis. Supplied as a high-purity, colorless to off-white solid, it is suitable for research-scale synthesis and formulation development; analytical documentation is available to support identity and purity.
  • High purity: 99.73% typical, suitable for synthetic use.
  • Molecular weight 180.20 g·mol-1 and formula C10H12O3 for reaction planning.
  • Available in laboratory pack sizes including 25 g for small-scale work.
  • Supplied with COA, H NMR, and RP-HPLC to confirm identity and purity.
  • Useful intermediate for pharmaceutical and fine-chemical syntheses.
Catalog No. 50-003-03523 Supplier Medchemexpress LLC Supplier No. 25GHYW027038
Ethyl mandelate is the ethyl ester of mandelic acid used as a pharmaceutical intermediate and building block in organic synthesis. Supplied as a high-purity, colorless to off-white solid, it is suitable for research-scale synthesis and formulation development; analytical documentation is available to support identity and purity.
  • High purity: 99.73% typical, suitable for synthetic use.
  • Molecular weight 180.20 g·mol-1 and formula C10H12O3 for reaction planning.
  • Available in laboratory pack sizes including 25 g for small-scale work.
  • Supplied with COA, H NMR, and RP-HPLC to confirm identity and purity.
  • Useful intermediate for pharmaceutical and fine-chemical syntheses.