Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetic acid, α-(methoxyimino)-2-[(2-methylphenoxy)methyl]-, methyl ester, (αE)- | 143390-89-0 | MFCD00871777 | 99.8% | 313.35 g/mol | C18H19NO4 | 25MG
SDP

Catalog No. 5000236522
Encompass_Preferred

Kresoxim-methyl is a strobilurin-class fungicide supplied as a research standard for biochemical and agronomic studies. It inhibits mitochondrial complex III (cytochrome bc1) and is provided as a high-purity solid suitable for analytical, assay, and reference uses.

  • Purity: 99.8% (HPLC).
  • Appearance: white to off-white solid.
  • Molecular formula: C18H19NO4.
  • Molecular weight: 313.35 g/mol.
  • Storage: powder stable at -20°C for long-term; in solvent store at -80°C for extended periods.
  • Available pack sizes: 5 mg-100 mg, suitable for analytical workflows.
  • Intended use: reference standard for analytical and agronomic research.

Catalog No. 50-002-36522 Supplier Medchemexpress LLC Supplier No. HY12577625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Kresoxim-methyl is a strobilurin-class fungicide supplied as a research standard for biochemical and agronomic studies. It inhibits mitochondrial complex III (cytochrome bc1) and is provided as a high-purity solid suitable for analytical, assay, and reference uses.

  • Purity: 99.8% (HPLC).
  • Appearance: white to off-white solid.
  • Molecular formula: C18H19NO4.
  • Molecular weight: 313.35 g/mol.
  • Storage: powder stable at -20°C for long-term; in solvent store at -80°C for extended periods.
  • Available pack sizes: 5 mg-100 mg, suitable for analytical workflows.
  • Intended use: reference standard for analytical and agronomic research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.