Learn More
Medchemexpress LLC Benzeneacetic acid, α-methyl-3-phenoxy- | 29679-58-1 | 99.7% | 100 MG
SDP
Catalog No. 5000413248
Encompass_Preferred
Fenoprofen is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits cyclooxygenase (COX). It acts as a positive allosteric modulator (PAM) of melanocortin receptors (MCRs) and increases ERK1/2 activation in HEK293T cells. It possesses anti-arthritic activities and is used for studying rheumatoid arthritis and osteoarthritis.
  • Inhibits cyclooxygenase.
  • Acts as a positive allosteric modulator of melanocortin receptors.
  • Increases ERK1/2 activation in HEK293T cells.
  • Has anti-arthritic activities.
  • Can be used for the study of rheumatoid arthritis and osteoarthritis.
  • Stimulates ERK1/2 activation at MC3-5R in a biphasic manner.
  • Induces biased signaling at MC3-5R, selectively activating ERK1/2 cascade.
  • Presents anti-arthritic actions on cartilage integrity and synovitis.
Catalog No. 50-004-13248 Supplier Medchemexpress LLC Supplier No. HYB1456A100MG
Fenoprofen is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits cyclooxygenase (COX). It acts as a positive allosteric modulator (PAM) of melanocortin receptors (MCRs) and increases ERK1/2 activation in HEK293T cells. It possesses anti-arthritic activities and is used for studying rheumatoid arthritis and osteoarthritis.
  • Inhibits cyclooxygenase.
  • Acts as a positive allosteric modulator of melanocortin receptors.
  • Increases ERK1/2 activation in HEK293T cells.
  • Has anti-arthritic activities.
  • Can be used for the study of rheumatoid arthritis and osteoarthritis.
  • Stimulates ERK1/2 activation at MC3-5R in a biphasic manner.
  • Induces biased signaling at MC3-5R, selectively activating ERK1/2 cascade.
  • Presents anti-arthritic actions on cartilage integrity and synovitis.