Learn More
Medchemexpress LLC Benzeneacetic acid, α-methyl-3-phenoxy- | 29679-58-1 | 99.7% | 250 MG
SDP
Catalog No. 5000414489
Encompass_Preferred
Fenoprofen is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits cyclooxygenase (COX). It acts as a positive allosteric modulator of melanocortin receptors (MCRs), increasing ERK1/2 activation in HEK293T cells. It is utilized in the study of rheumatoid arthritis and osteoarthritis.
  • Inhibits cyclooxygenase (COX)
  • Acts as a positive allosteric modulator of melanocortin receptors
  • Increases ERK1/2 activation
  • Exhibits anti-arthritic activities
  • Used in research for rheumatoid arthritis
  • Used in research for osteoarthritis
  • For research use only
Catalog No. 50-004-14489 Supplier Medchemexpress LLC Supplier No. 250MGHYB1456A
Fenoprofen is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits cyclooxygenase (COX). It acts as a positive allosteric modulator of melanocortin receptors (MCRs), increasing ERK1/2 activation in HEK293T cells. It is utilized in the study of rheumatoid arthritis and osteoarthritis.
  • Inhibits cyclooxygenase (COX)
  • Acts as a positive allosteric modulator of melanocortin receptors
  • Increases ERK1/2 activation
  • Exhibits anti-arthritic activities
  • Used in research for rheumatoid arthritis
  • Used in research for osteoarthritis
  • For research use only