Learn More
Medchemexpress LLC Benzenecarboselenoic acid, 4-chloro-, Se-[(2,2-dimethyl-1,3-dioxolan-4-yl)methyl] ester | 2098850-81-6 | 98.9% | 333.67 g/mol | C13H15ClO3Se | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1397035MG
Encompass_Preferred
Se-DMC is a selenium-containing small-molecule research compound (CAS 2098850-81-6) reported to attenuate Complete Freund's Adjuvant (CFA)-induced inflammatory responses, nociception, and neurobehavioral deficits in mice. Supplied for laboratory research use only; not for human or veterinary use. Molecular formula C13H15ClO3Se; molecular weight 333.67 g/mol; reported purity 98.88%.
  • Selenium-containing small-molecule with anti-inflammatory activity.
  • Demonstrated efficacy in CFA-induced inflammation and nociception models in mice.
  • High reported purity suitable for biochemical assays.
  • Soluble in DMSO for in vitro applications.
  • Provided for research use only; not for clinical use.
Catalog No. 50-002-22715