Learn More
Medchemexpress LLC Benzenecarboximidamide, hydrochloride, hydrate | 206752-36-5 | 99.6% | C7H8N2·ClH·xH2O | 5 G
SDP
Catalog No. 5000208369
Encompass_Preferred
Benzamidine hydrochloride hydrate is a reversible competitive inhibitor of trypsin-like serine proteases used in enzymatic assays and protease inhibition studies. It is supplied as a white to light-yellow solid with high purity for biochemical research.
  • Reversible competitive inhibitor of trypsin-like serine proteases.
  • High purity: 99.58%.
  • White to light-yellow solid physical form.
  • Available in common lab pack sizes, including 5 g.
  • Intended for research use in enzymology and protease inhibition assays.
Catalog No. 50-002-08369 Supplier Medchemexpress LLC Supplier No. HYW0879375G
Benzamidine hydrochloride hydrate is a reversible competitive inhibitor of trypsin-like serine proteases used in enzymatic assays and protease inhibition studies. It is supplied as a white to light-yellow solid with high purity for biochemical research.
  • Reversible competitive inhibitor of trypsin-like serine proteases.
  • High purity: 99.58%.
  • White to light-yellow solid physical form.
  • Available in common lab pack sizes, including 5 g.
  • Intended for research use in enzymology and protease inhibition assays.