Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenehexachloride, MP Biomedicals

Catalog No. ICN204985 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204985 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204985 Supplier MP Biomedicals, Inc Supplier No. 0520498580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58-89-9
Molecular Formula C6H6Cl6
Molecular Weight (g/mol) 290.81
MDL Number MFCD00135947
InChI Key JLYXXMFPNIAWKQ-UHFFFAOYSA-N
Synonym lindane, beta-hch, gamma-hch, gamma-bhc, hexachlorane, alpha-hch, kwell, hexicide, beta-bhc, beta-lindane
PubChem CID 727
ChEBI CHEBI:24536
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
SMILES ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl

Specifications

Quantity 100 g
Formula Weight 290.83
Chemical Name or Material Benzene Hexachloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.