Learn More
Medchemexpress LLC Benzenemethanaminium, N-[4-[[4-[(4-ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, inner salt, sodium salt (1:1) | 6104-59-2 | 95.0% | 50 G
SDP
Supplier:  Medchemexpress LLC HYD023250G
Encompass_Preferred
Brilliant blue R250 (Brilliant Blue R) is an anionic dye commonly used as the most popular stain to detect proteins resolved in SDS-PAGE gels. It is intended for research use only.
  • Anionic dye
  • Used to detect proteins resolved in SDS-PAGE gels
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-004-09764