Learn More
Medchemexpress LLC Benzenemethanol, α-[[([1,1'-biphenyl]-4-ylmethyl)amino]methyl]-3-hydroxy | 775294-71-8 | 99.8% | 100 MG
SDP
Catalog No. 5000230578
Encompass_Preferred
AC-73 is a small-molecule inhibitor of cluster of differentiation 147 (CD147) that disrupts CD147 dimerization and inhibits cell motility, invasion, and proliferation. It is supplied for preclinical research use in solid and solution formats for biochemical and cell-based assays.
  • First specific orally active CD147 inhibitor.
  • Disrupts CD147 dimerization to inhibit motility and invasion.
  • Exhibits anti-proliferative and autophagy-inducing activity in leukemic cells.
  • High purity suitable for research use (purity 99.8%).
  • Available as solid powder and as 10 mM solutions in DMSO.
  • Offered in multiple package sizes, including 100 mg.
  • Intended for pathway inhibition, migration, and invasion studies.
Catalog No. 50-002-30578 Supplier Medchemexpress LLC Supplier No. HY122214100MG
AC-73 is a small-molecule inhibitor of cluster of differentiation 147 (CD147) that disrupts CD147 dimerization and inhibits cell motility, invasion, and proliferation. It is supplied for preclinical research use in solid and solution formats for biochemical and cell-based assays.
  • First specific orally active CD147 inhibitor.
  • Disrupts CD147 dimerization to inhibit motility and invasion.
  • Exhibits anti-proliferative and autophagy-inducing activity in leukemic cells.
  • High purity suitable for research use (purity 99.8%).
  • Available as solid powder and as 10 mM solutions in DMSO.
  • Offered in multiple package sizes, including 100 mg.
  • Intended for pathway inhibition, migration, and invasion studies.