Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenemethanol, 3-[3-[bis(1-methylethyl)amino]-1-phenylpropyl]-4-hydroxy- | 200801-70-3 | 99.05% | C22H31NO2 | 1 ML
SDP

Catalog No. 5000434436
Encompass_Preferred

(Rac)-5-Hydroxymethyl Tolterodine is an active metabolite of Tolterodine and a mAChR antagonist, targeting M1, M2, M3, M4, and M5 receptors with Ki values ranging from 2.3 nM to 2.9 nM. It is suitable for overactive bladder research.

  • Active metabolite of Tolterodine
  • mAChR antagonist
  • Targets M1, M2, M3, M4, and M5 receptors
  • Can be used for overactive bladder research
  • Available as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • In vivo solubility protocols are available

Catalog No. 50-004-34436 Supplier Medchemexpress LLC Supplier No. HY7657010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-5-Hydroxymethyl Tolterodine is an active metabolite of Tolterodine and a mAChR antagonist, targeting M1, M2, M3, M4, and M5 receptors with Ki values ranging from 2.3 nM to 2.9 nM. It is suitable for overactive bladder research.

  • Active metabolite of Tolterodine
  • mAChR antagonist
  • Targets M1, M2, M3, M4, and M5 receptors
  • Can be used for overactive bladder research
  • Available as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • In vivo solubility protocols are available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.