Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenemethanol, 3-hydroxy- | 620-24-6 | 99.9% | C7H8O2 | 25 G
SDP

Catalog No. 5000357936
Encompass_Preferred

KSD 2405 is an endogenous metabolite characterized by its molecular formula C7H8O2 and molecular weight of 124.14. This compound is a light brown to brown solid with a high purity of 99.9%.

  • CAS number: 620-24-6
  • Purity (HPLC): 99.9%
  • Molecular formula: C7H8O2
  • Molecular weight: 124.14
  • Appearance: Light brown to brown solid
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 2 years or -20°C for 1 year

Catalog No. 50-003-57936 Supplier Medchemexpress LLC Supplier No. 25GHY78446
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KSD 2405 is an endogenous metabolite characterized by its molecular formula C7H8O2 and molecular weight of 124.14. This compound is a light brown to brown solid with a high purity of 99.9%.

  • CAS number: 620-24-6
  • Purity (HPLC): 99.9%
  • Molecular formula: C7H8O2
  • Molecular weight: 124.14
  • Appearance: Light brown to brown solid
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.