Learn More
Medchemexpress LLC Benzenemethanol, 4-amino-3,5-dichloro-α-[[[1,1-di(methyl-d3)ethyl-2,2,2-d3]amino]methyl]- | 129138-58-5 | ≥99.0% | 5 MG
SDP
Catalog No. 5000411662
Encompass_Preferred
Clenbuterol-d9 is a deuterium labeled form of Clenbuterol. Clenbuterol (NAB-365) is a β2-adrenergic receptor agonist with an EC50 of 31.9 nM. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11662 Supplier Medchemexpress LLC Supplier No. HYB1615S5MG
Clenbuterol-d9 is a deuterium labeled form of Clenbuterol. Clenbuterol (NAB-365) is a β2-adrenergic receptor agonist with an EC50 of 31.9 nM. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.