Learn More
Medchemexpress LLC Benzenemethanol, 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1-acetate | 2142562-01-2 | 99.6% | 294.13 | 100 MG
SDP
Catalog No. 5000403854
Encompass_Preferred
5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl acetate is a chemical compound for research use only. It appears as a white to off-white solid with a molecular weight of 294.13 and a purity of 99.62%.
  • Appears as a white to off-white solid
  • High purity of 99.62%
  • Molecular formula C15H20BFO4
  • Consistent with 1H NMR spectrum and LCMS
  • Suitable for sealed storage at 4°C, or in solvent at -80°C or -20°C for extended periods
Catalog No. 50-004-03854 Supplier Medchemexpress LLC Supplier No. HY43489100MG
Add to Cart
Add to Cart
5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl acetate is a chemical compound for research use only. It appears as a white to off-white solid with a molecular weight of 294.13 and a purity of 99.62%.
  • Appears as a white to off-white solid
  • High purity of 99.62%
  • Molecular formula C15H20BFO4
  • Consistent with 1H NMR spectrum and LCMS
  • Suitable for sealed storage at 4°C, or in solvent at -80°C or -20°C for extended periods