Learn More
Medchemexpress LLC Benzenemethanol,a,a-dimethyl-4-[(1Z)-2-[1-[4-(1-methylethoxy)phenyl]-1H-benzimidazol-5-yl]ethen | 1450929-77-7 | 99.1% | 412.52 | 50 MG
SDP
Catalog No. 5000436103
Encompass_Preferred
LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38. It has EC50s of <1 μM for Lassa, Machupo, and Junin viruses, and 1-10 μM for VSVg virus. It is a small-molecule viral entry inhibitor that targets the arenavirus envelope glycoprotein (GP).
  • Exhibits potent antiviral activity against a broad array of hemorrhagic fever arenaviruses.
  • Inhibits Lassa GP-pseudotyped lentivirus with an IC50 of 0.1-0.3 nM.
  • Protects mice from a lethal challenge with Tacaribe virus and dramatically reduces viral titers in plasma, spleen, and liver.
  • An increase in survival is also observed when the first dose is delayed by 1, 2, or 3 days after infection in mice.
  • Effective as a post-exposure therapeutic.
Catalog No. 50-004-36103 Supplier Medchemexpress LLC Supplier No. HY11276250MG
LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38. It has EC50s of <1 μM for Lassa, Machupo, and Junin viruses, and 1-10 μM for VSVg virus. It is a small-molecule viral entry inhibitor that targets the arenavirus envelope glycoprotein (GP).
  • Exhibits potent antiviral activity against a broad array of hemorrhagic fever arenaviruses.
  • Inhibits Lassa GP-pseudotyped lentivirus with an IC50 of 0.1-0.3 nM.
  • Protects mice from a lethal challenge with Tacaribe virus and dramatically reduces viral titers in plasma, spleen, and liver.
  • An increase in survival is also observed when the first dose is delayed by 1, 2, or 3 days after infection in mice.
  • Effective as a post-exposure therapeutic.